missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%

Catalog No. AAH51108MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51108MD 250 mg
AAH5110803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51108MD Supplier Thermo Scientific Chemicals Supplier No. H51108MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77145-39-2 | C10H16ClN3 | 213.709 g/mol

4-Amino-1-(2-pyridyl)piperidine hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-(2-pyridyl)piperidine hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 77145-39-2
Molecular Formula C10H16ClN3
Molecular Weight (g/mol) 213.709
MDL Number MFCD09607833
InChI Key HNICPQFPDAEZKW-UHFFFAOYSA-N
Synonym 1-pyridin-2-yl piperidin-4-amine hydrochloride, 4-amino-1-2-pyridyl piperidine hydrochloride, 1-2-pyridyl-4-piperidylamine, chloride, 1-2-pyridyl-4-piperidinamine hydrochloride, 1-pyridin-2-yl piperidin-4-amine-hydrogen chloride 1/1, 1-pyridin-2-yl-piperazine hydrochloride; 1-2-pyridyl-4-piperidinamine hydrochloride
PubChem CID 45786931
IUPAC Name 1-pyridin-2-ylpiperidin-4-amine;hydrochloride
SMILES C1CN(CCC1N)C2=CC=CC=N2.Cl

Specifications

Quantity 250 mg
Formula Weight 213.71
Percent Purity 97%
Chemical Name or Material 4-Amino-1-(2-pyridyl)piperidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only