missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-[2-(4-methoxyphenyl)ethyl]piperidine 98.0+%, TCI America™

Catalog No. A161825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A161825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A161825G Supplier TCI America Supplier No. A161825G

Chemical Identifiers

CAS 85098-70-0
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.34
MDL Number MFCD01631147
InChI Key HTGMIBQQTLUEKV-UHFFFAOYSA-N
Synonym 4-Amino-1-(4-methoxyphenethyl)piperidine, 1-(4-Methoxyphenethyl)-4-aminopiperidine, 1-[2-(4-Methoxyphenyl)ethyl]-4-aminopiperidine
PubChem CID 3020332
IUPAC Name 1-[2-(4-methoxyphenyl)ethyl]piperidin-4-amine
SMILES COC1=CC=C(CCN2CCC(N)CC2)C=C1

Specifications

Quantity 25 g
Formula Weight 234.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1-[2-(4-methoxyphenyl)ethyl]piperidine