missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,2,6,6-pentamethylpiperidine, 99%

Catalog No. p-3586585
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC335940010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC335940010 Supplier Thermo Scientific Chemicals Supplier No. 335940010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40327-96-6 | C10H24N2 | 172.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40327-96-6
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.32
MDL Number MFCD01861832
InChI Key CGXOAAMIQPDTPE-UHFFFAOYSA-P
Synonym 4-amino-1,2,2,6,6-pentamethylpiperidine, 1,2,2,6,6-pentamethyl-4-aminopiperidine, 1,2,2,6,6-pentamethyl-4-piperidinamine, 1,2,2,6,6-pentamethyl-4-piperidylamine, 4-ammo-1,2,2,6,6-pentamethylpiperidine, 4-piperidinamine,1,2,2,6,6-pentamethyl, 1,2,-2,6,6-pentamethyl-4-aminopiperidine, 1,2,2,6,6,-pentamethyl-4-aminopiperidine, 4-amino-1,2,2,6,6-pentamethyl-piperidine, 4-amino-1,2,2,6,6-pentamethyl piperidine, 97+%
PubChem CID 693792
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-amine
SMILES C[NH+]1C(C)(C)CC([NH3+])CC1(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 215°C to 218°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4795 to 1.4815
Quantity 1 g
Beilstein 22, II, 321
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 170.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Amino-1,2,2,6,6-pentamethylpiperidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only