accessibility menu, dialog, popup

UserName

CAS RN 40327-96-6

IUPAC Name: 4-azaniumyl-1,2,2,6,6-pentamethylpiperidin-1-ium
Synonyms: 4-ammonio-1,2,2,6,6-pentamethylpiperidin-1-ium
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: CGXOAAMIQPDTPE-UHFFFAOYSA-P
SMILES: C[NH+]1C(C)(C)CC([NH3+])CC1(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results