missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-alpha-Cumylphenol 98.0+%, TCI America™
$32.72 - $92.41
Chemical Identifiers
| CAS | 599-64-4 |
|---|---|
| Molecular Formula | C15H16O |
| Molecular Weight (g/mol) | 212.292 |
| MDL Number | MFCD00002365 |
| InChI Key | QBDSZLJBMIMQRS-UHFFFAOYSA-N |
| Synonym | 4-cumylphenol, p-cumylphenol, 4-2-phenylpropan-2-yl phenol, p-cumyl phenol, phenol, 4-1-methyl-1-phenylethyl, 4-1-methyl-1-phenylethyl phenol, 4-alpha-cumylphenol, 4-dimethylphenylmethyl phenol, 4-hydroxydiphenyldimethylmethane, 2-phenyl-2-4-hydroxyphenyl propane |
| PubChem CID | 11742 |
| ChEBI | CHEBI:35092 |
| IUPAC Name | 4-(2-phenylpropan-2-yl)phenol |
| SMILES | CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O |
Chemical Identifiers
| 599-64-4 | |
| 212.292 | |
| QBDSZLJBMIMQRS-UHFFFAOYSA-N | |
| 11742 | |
| 4-(2-phenylpropan-2-yl)phenol |
| C15H16O | |
| MFCD00002365 | |
| 4-cumylphenol, p-cumylphenol, 4-2-phenylpropan-2-yl phenol, p-cumyl phenol, phenol, 4-1-methyl-1-phenylethyl, 4-1-methyl-1-phenylethyl phenol, 4-alpha-cumylphenol, 4-dimethylphenylmethyl phenol, 4-hydroxydiphenyldimethylmethane, 2-phenyl-2-4-hydroxyphenyl propane | |
| CHEBI:35092 | |
| CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O |
Specifications
| 599-64-4 | |
| White-Yellow | |
| C15H16O | |
| 25 g | |
| 4-cumylphenol, p-cumylphenol, 4-2-phenylpropan-2-yl phenol, p-cumyl phenol, phenol, 4-1-methyl-1-phenylethyl, 4-1-methyl-1-phenylethyl phenol, 4-alpha-cumylphenol, 4-dimethylphenylmethyl phenol, 4-hydroxydiphenyldimethylmethane, 2-phenyl-2-4-hydroxyphenyl propane | |
| CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O | |
| 212.292 | |
| CHEBI:35092 | |
| ≥98.0% (GC) | |
| 4-alpha-Cumylphenol |
| 73°C | |
| 187°C | |
| MFCD00002365 | |
| 3077 | |
| QBDSZLJBMIMQRS-UHFFFAOYSA-N | |
| 4-(2-phenylpropan-2-yl)phenol | |
| 11742 | |
| 212.29 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0122
RTECSNumber : SL1942450
TSCA : Yes