missing translation for 'onlineSavingsMsg'
Learn More

4-alpha-Cumylphenol 98.0+%, TCI America™

Catalog No. C0684500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0684500G 500 g
C068425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0684500G Supplier TCI America Supplier No. C0684500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 599-64-4
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00002365
InChI Key QBDSZLJBMIMQRS-UHFFFAOYSA-N
Synonym 4-cumylphenol, p-cumylphenol, 4-2-phenylpropan-2-yl phenol, p-cumyl phenol, phenol, 4-1-methyl-1-phenylethyl, 4-1-methyl-1-phenylethyl phenol, 4-alpha-cumylphenol, 4-dimethylphenylmethyl phenol, 4-hydroxydiphenyldimethylmethane, 2-phenyl-2-4-hydroxyphenyl propane
PubChem CID 11742
ChEBI CHEBI:35092
IUPAC Name 4-(2-phenylpropan-2-yl)phenol
SMILES CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 187°C
Quantity 500 g
UN Number 3077
Formula Weight 212.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-alpha-Cumylphenol
EINECSNumber (4)-0122
RTECSNumber SL1942450
TSCA Yes