missing translation for 'onlineSavingsMsg'
Learn More

4-Allylmorpholine 97.0+%, TCI America™

Catalog No. p-7117047
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
A07565ML 5 mL
A075625ML 25 mL
2 options

Catalog No. A07565ML

Supplier: TCI America A07565ML

Only null left

Chemical Identifiers

CAS 696-57-1
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00038034
InChI Key SUSANAYXICMXBL-UHFFFAOYSA-N
Synonym 4-allylmorpholine, n-allylmorpholine, allylmorpholine, n-allyl morpholine, 4-prop-2-en-1-yl morpholine, allyl morpholine, n-allymorpholine, 4-allyl-morpholine, 4-allylmorpholine #, morpholine, 4-allyl
PubChem CID 136522
IUPAC Name 4-prop-2-enylmorpholine
SMILES C=CCN1CCOCC1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 mL
UN Number 1993
Formula Weight 127.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Allylmorpholine