accessibility menu, dialog, popup

UserName

CH2NO

CAS RN 696-57-1

IUPAC Name: 4-(prop-2-en-1-yl)morpholine
Synonyms: 4-(prop-2-en-1-yl)morpholine
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: SUSANAYXICMXBL-UHFFFAOYSA-N
SMILES: C=CCN1CCOCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results