missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Allyl-2-methoxyphenoxy)acetic Acid 98.0+%, TCI America™
$55.92 - $279.60
Chemical Identifiers
| CAS | 6331-61-9 |
|---|---|
| Molecular Formula | C12H14O4 |
| Molecular Weight (g/mol) | 222.24 |
| MDL Number | MFCD00017594 |
| InChI Key | FQBPCNCFOCPCJP-UHFFFAOYSA-N |
| PubChem CID | 235909 |
| IUPAC Name | 2-(2-methoxy-4-prop-2-enylphenoxy)acetic acid |
| SMILES | COC1=C(C=CC(=C1)CC=C)OCC(=O)O |
Chemical Identifiers
| 6331-61-9 | |
| 222.24 | |
| FQBPCNCFOCPCJP-UHFFFAOYSA-N | |
| 2-(2-methoxy-4-prop-2-enylphenoxy)acetic acid |
| C12H14O4 | |
| MFCD00017594 | |
| 235909 | |
| COC1=C(C=CC(=C1)CC=C)OCC(=O)O |
Specifications
| 6331-61-9 | |
| C12H14O4 | |
| 1 g | |
| COC1=C(C=CC(=C1)CC=C)OCC(=O)O | |
| 222.24 | |
| 222.24 | |
| Crystalline Powder |
| 101°C | |
| MFCD00017594 | |
| FQBPCNCFOCPCJP-UHFFFAOYSA-N | |
| 2-(2-methoxy-4-prop-2-enylphenoxy)acetic acid | |
| 235909 | |
| ≥98.0% (GC,T) | |
| (4-Allyl-2-methoxyphenoxy)acetic Acid |
Safety and Handling
RTECSNumber : AI8967340
TSCA : No