missing translation for 'onlineSavingsMsg'
Learn More

(4-Allyl-2-methoxyphenoxy)acetic Acid 98.0+%, TCI America™

Catalog No. A30375G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30375G 5 g
A30371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30375G Supplier TCI America Supplier No. A30375G
Only null left

Chemical Identifiers

CAS 6331-61-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00017594
InChI Key FQBPCNCFOCPCJP-UHFFFAOYSA-N
PubChem CID 235909
IUPAC Name 2-(2-methoxy-4-prop-2-enylphenoxy)acetic acid
SMILES COC1=C(C=CC(=C1)CC=C)OCC(=O)O

Specifications

Melting Point 101°C
Quantity 5 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Allyl-2-methoxyphenoxy)acetic Acid
RTECSNumber AI8967340
TSCA No