missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Allyl-2-methoxyphenoxy)acetic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 101°C |
| Quantity | 1 g |
| Formula Weight | 222.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (4-Allyl-2-methoxyphenoxy)acetic Acid |
Safety and Handling
| RTECSNumber | AI8967340 |
| TSCA | No |