missing translation for 'onlineSavingsMsg'
Learn More

4-Allyl-2,6-dimethoxyphenol, 98%

Catalog No. AAB2198106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2198106 5 g
AAB2198103 1 g
AAB2198114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2198106 Supplier Thermo Scientific Chemicals Supplier No. B2198106

CAS: 6627-88-9 | C11H14O3 | 194.23 g/mol

4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.

Solubility
Soluble in fats. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 6627-88-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008655
InChI Key FWMPKHMKIJDEMJ-UHFFFAOYSA-N
Synonym 4-allyl-2,6-dimethoxyphenol, methoxyeugenol, 4-allylsyringol, phenol, 2,6-dimethoxy-4-2-propenyl, 2,6-dimethoxychavicol, phenol, 4-allyl-2,6-dimethoxy, unii-8vf00ywp89, 2,6-dimethoxy-4-allylphenol, 2,6-dimethoxy-4-prop-2-en-1-yl phenol, 4-hydroxy-3,5-dimethoxyallylbenzene
PubChem CID 226486
ChEBI CHEBI:86562
IUPAC Name 2,6-dimethoxy-4-prop-2-enylphenol
SMILES COC1=CC(CC=C)=CC(OC)=C1O

Specifications

Density 1.092
Boiling Point 168°C to 169°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 5 g
Solubility Information Soluble in fats. Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 4-Allyl-2,6-dimethoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-600-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only