missing translation for 'onlineSavingsMsg'
Learn More

4-Allyl-1,6-heptadien-4-ol, 99%

CAS: 10202-75-2 | C10H16O | 152.237 g/mol

Supplier:  Thermo Scientific Chemicals B2070309

 View more versions of this product

Catalog No. AAB2070309


Only null left
Explore available promotions

Description

Description

A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Allyl-1,6-heptadien-4-ol
C10H16O
10 g
Soluble in water.
C=CCC(CC=C)(CC=C)O
152.237
152.24
10202-75-2
MFCD00039829
4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl
SUXQWOWVXDXQSE-UHFFFAOYSA-N
4-prop-2-enylhepta-1,6-dien-4-ol
139107
99%

Chemical Identifiers

10202-75-2
152.237
SUXQWOWVXDXQSE-UHFFFAOYSA-N
139107
C=CCC(CC=C)(CC=C)O
C10H16O
MFCD00039829
4-allyl-1,6-heptadien-4-ol, triallylcarbinol, 4-allylhepta-1,6-dien-4-ol, 4-allyl-1,6-heptadiene-4-ol, 1,6-heptadien-4-ol, 4-2-propenyl, 4-prop-2-en-1-yl hepta-1,6-dien-4-ol, triallyl carbinol, acmc-1byg9, 4-allyl-hepta-1,6-dien-4-ol, 1,6-heptadien-4-ol,4-2-propen-1-yl
4-prop-2-enylhepta-1,6-dien-4-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only