CAS RN 10202-75-2
IUPAC Name:
4-(prop-2-en-1-yl)hepta-1,6-dien-4-ol
Synonyms:
4-(prop-2-en-1-yl)hepta-1,6-dien-4-ol
Molecular Weight (g/mol):
152.24
Molecular Formula:
C10H16O
InChi Key:
SUXQWOWVXDXQSE-UHFFFAOYSA-N
SMILES:
OC(CC=C)(CC=C)CC=C
1
–
1
of
1
results
1
Promotions Available
4-Allyl-1,6-heptadien-4-ol, 99%
4-Allyl-1,6-heptadien-4-ol, 99%
CAS: 10202-75-2 Molecular Formula: C10H16O Molecular Weight (g/mol): 152.237 MDL Number: MFCD00039829 InChI Key: SUXQWOWVXDXQSE-UHFFFAOYSA-N Synonym: 4-allyl-1,6-heptadien-4-ol,triallylcarbinol,4-allylhepta-1,6-dien-4-ol,4-allyl-1,6-heptadiene-4-ol,1,6-heptadien-4-ol, 4-2-propenyl,4-prop-2-en-1-yl hepta-1,6-dien-4-ol,triallyl carbinol,acmc-1byg9,4-allyl-hepta-1,6-dien-4-ol,1,6-heptadien-4-ol,4-2-propen-1-yl PubChem CID: 139107 IUPAC Name: 4-prop-2-enylhepta-1,6-dien-4-ol SMILES: C=CCC(CC=C)(CC=C)O