missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyltetrahydropyran, 97%

Catalog No. p-7055839
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64522MD 250 mg
AAH6452203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64522MD Supplier Thermo Scientific Chemicals Supplier No. H64522MD
Only null left

CAS: 137052-08-5 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137052-08-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD08704647
InChI Key VNMXIOWPBADSIC-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-4-yl ethanone, 1-oxan-4-yl ethan-1-one, 1-oxan-4-yl ethanone, 4-acetyltetrahydropyran, 1-tetrahydro-2h-pyran-4-ylethanone, 4-acetyltetrahydro-4h-pyran, 4-acetyltetrahydro-2h-pyran, 1-tetrahydropyran-4-ylethanone, 1-tetrahydro-pyran-4-yl-ethanone, 1-tetrahydro-2h-pyran-4-yl ethan-1-one
PubChem CID 9877365
IUPAC Name 1-(oxan-4-yl)ethanone
SMILES CC(=O)C1CCOCC1

Specifications

Color Colorless to Yellow
Density 1.024 g/mL
Boiling Point 90°C to 94°C (15 mmHg)
Refractive Index 1.453
Quantity 250 mg
Beilstein 109587
Formula Weight 128.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Acetyltetrahydropyran
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only