missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylpyridine, 98%

CAS: 1122-54-9 | C7H7NO | 121.14 g/mol

$48.98 - $211.18

Chemical Identifiers

CAS 1122-54-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00006433
InChI Key WMQUKDQWMMOHSA-UHFFFAOYSA-N
Synonym 4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
PubChem CID 14282
IUPAC Name 1-pyridin-4-ylethanone
SMILES CC(=O)C1=CC=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102700100
SDS View Documents
Thermo Scientific Chemicals
102700100
10 g Glass bottle
Each for $48.98
Only null left
 
AC102701000
SDS View Documents
Thermo Scientific Chemicals
102701000
100 g Glass bottle
Each for $211.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1122-54-9
121.14
WMQUKDQWMMOHSA-UHFFFAOYSA-N
14282
CC(=O)C1=CC=NC=C1
C7H7NO
MFCD00006433
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
1-pyridin-4-ylethanone

Specifications

1122-54-9
100.0
Brown to Yellow
212.0°C
Authentic
Glass bottle
1.5270 to 1.5310
MFCD00006433
21, 279
4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
WMQUKDQWMMOHSA-UHFFFAOYSA-N
1-pyridin-4-ylethanone
14282
98%
4-Acetylpyridine
97.5
13.0°C to 16.0°C
1.0900g/mL
104°C
97.5% min. (GC)
C7H7NO
(C5H4N)COCH3
10 g
1.09
Solubility in water: insoluble
CC(=O)C1=CC=NC=C1
121.14
121.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

EINECSNumber : 214-350-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only