missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylpyridine, 98%

Catalog No. AC102700100
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102700100 10 g Glass bottle
AC102701000 100 g Glass bottle
AC102705000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102700100 Supplier Thermo Scientific Chemicals Supplier No. 102700100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1122-54-9 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1122-54-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00006433
InChI Key WMQUKDQWMMOHSA-UHFFFAOYSA-N
Synonym 4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
PubChem CID 14282
IUPAC Name 1-pyridin-4-ylethanone
SMILES CC(=O)C1=CC=NC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 13.0°C to 16.0°C
Color Brown to Yellow
Density 1.0900g/mL
Boiling Point 212.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5270 to 1.5310
Linear Formula (C5H4N)COCH3
Quantity 10 g
Beilstein 21, 279
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Acetylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 214-350-9

RUO – Research Use Only