missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T17095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17095G Supplier TCI America Supplier No. T17095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109613-00-5
Molecular Formula C9H7F3O4S
Molecular Weight (g/mol) 268.206
MDL Number MFCD00191706
InChI Key RUMMIUOXQGFCEP-UHFFFAOYSA-N
Synonym 4-Acetylphenyl Triflate, Trifluoromethanesulfonic Acid 4-Acetylphenyl Ester
PubChem CID 2769355
IUPAC Name (4-acetylphenyl) trifluoromethanesulfonate
SMILES CC(=O)C1=CC=C(C=C1)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 268.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Acetylphenyl Trifluoromethanesulfonate