accessibility menu, dialog, popup

UserName

CAS RN 109613-00-5

IUPAC Name: 4-acetylphenyl trifluoromethanesulfonate
Synonyms: 4-acetylphenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 268.21
Molecular Formula: C9H7F3O4S
InChi Key: RUMMIUOXQGFCEP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results