missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Acetylphenoxy)acetic Acid 98.0+%, TCI America™
Supplier: TCI America A218225G
Specifications
| (4-Acetylphenoxy)acetic Acid | |
| 174°C | |
| C10H10O4 | |
| 25 g | |
| KMXZEXUYXUMHEQ-UHFFFAOYSA-N | |
| 2-(4-acetylphenoxy)acetic acid | |
| 74655 | |
| ≥98.0% (GC,T) |
| 1878-81-5 | |
| Yellow | |
| MFCD00014363 | |
| 4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy | |
| CC(=O)C1=CC=C(OCC(O)=O)C=C1 | |
| 194.19 | |
| 194.19 | |
| Crystalline Powder |
Chemical Identifiers
| 1878-81-5 | |
| 194.19 | |
| KMXZEXUYXUMHEQ-UHFFFAOYSA-N | |
| 74655 | |
| CC(=O)C1=CC=C(OCC(O)=O)C=C1 |
| C10H10O4 | |
| MFCD00014363 | |
| 4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy | |
| 2-(4-acetylphenoxy)acetic acid |
Safety and Handling
TSCA : No