missing translation for 'onlineSavingsMsg'
Learn More

(4-Acetylphenoxy)acetic Acid 98.0+%, TCI America™

Catalog No. p-7117779
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21825G 5 g
A218225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21825G Supplier TCI America Supplier No. A21825G

Chemical Identifiers

CAS 1878-81-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00014363
InChI Key KMXZEXUYXUMHEQ-UHFFFAOYSA-N
Synonym 4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy
PubChem CID 74655
IUPAC Name 2-(4-acetylphenoxy)acetic acid
SMILES CC(=O)C1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 174°C
Color Yellow
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Acetylphenoxy)acetic Acid