missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylmorpholine 99.0+%, TCI America™
$68.97 - $445.51
Chemical Identifiers
| CAS | 1696-20-4 |
|---|---|
| Molecular Formula | C6H11NO2 |
| Molecular Weight (g/mol) | 129.159 |
| MDL Number | MFCD00006171 |
| InChI Key | KYWXRBNOYGGPIZ-UHFFFAOYSA-N |
| Synonym | 4-acetylmorpholine, n-acetylmorpholine, 1-morpholinoethanone, morpholine, 4-acetyl, n-acetylmorfolin, n-acetylmorfolin czech, ethanone, 1-4-morpholinyl, n-acetyl morpholine, 1-morpholin-4-yl ethan-1-one, acetylmorpholine |
| PubChem CID | 15543 |
| IUPAC Name | 1-morpholin-4-ylethanone |
| SMILES | CC(=O)N1CCOCC1 |
Chemical Identifiers
| 1696-20-4 | |
| 129.159 | |
| KYWXRBNOYGGPIZ-UHFFFAOYSA-N | |
| 15543 | |
| CC(=O)N1CCOCC1 |
| C6H11NO2 | |
| MFCD00006171 | |
| 4-acetylmorpholine, n-acetylmorpholine, 1-morpholinoethanone, morpholine, 4-acetyl, n-acetylmorfolin, n-acetylmorfolin czech, ethanone, 1-4-morpholinyl, n-acetyl morpholine, 1-morpholin-4-yl ethan-1-one, acetylmorpholine | |
| 1-morpholin-4-ylethanone |
Specifications
| 1696-20-4 | |
| Yellow | |
| C6H11NO2 | |
| 25 g | |
| KYWXRBNOYGGPIZ-UHFFFAOYSA-N | |
| 1-morpholin-4-ylethanone | |
| 15543 | |
| ≥99.0% (GC) | |
| 4-Acetylmorpholine |
| 14°C | |
| 208°C | |
| MFCD00006171 | |
| 4-acetylmorpholine, n-acetylmorpholine, 1-morpholinoethanone, morpholine, 4-acetyl, n-acetylmorfolin, n-acetylmorfolin czech, ethanone, 1-4-morpholinyl, n-acetyl morpholine, 1-morpholin-4-yl ethan-1-one, acetylmorpholine | |
| CC(=O)N1CCOCC1 | |
| 129.159 | |
| 129.16 | |
| Liquid |
Safety and Handling
RTECSNumber : QD7000000
TSCA : Yes