accessibility menu, dialog, popup

UserName

H3CONO

CAS RN 1696-20-4

IUPAC Name: 1-(morpholin-4-yl)ethan-1-one
Synonyms: morpholine, 4-acetyl-
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: KYWXRBNOYGGPIZ-UHFFFAOYSA-N
SMILES: CC(=O)N1CCOCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)