missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylmorpholine 99.0+%, TCI America™

Catalog No. A0102500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0102500G 500 g
A010225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0102500G Supplier TCI America Supplier No. A0102500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1696-20-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00006171
InChI Key KYWXRBNOYGGPIZ-UHFFFAOYSA-N
Synonym 4-acetylmorpholine, n-acetylmorpholine, 1-morpholinoethanone, morpholine, 4-acetyl, n-acetylmorfolin, n-acetylmorfolin czech, ethanone, 1-4-morpholinyl, n-acetyl morpholine, 1-morpholin-4-yl ethan-1-one, acetylmorpholine
PubChem CID 15543
IUPAC Name 1-morpholin-4-ylethanone
SMILES CC(=O)N1CCOCC1

Specifications

Melting Point 14°C
Color Yellow
Boiling Point 208°C
Quantity 500 g
Formula Weight 129.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Acetylmorpholine
RTECSNumber QD7000000
TSCA Yes