Learn More
4-Acetylbutyric acid, 97%
CAS: 3128-06-1 | C6H10O3 | 130.14 g/mol
Supplier: Thermo Scientific Chemicals L0397814
Description
4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.
Solubility
Slightly soluble in water, chloroform and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
| 4-Acetylbutyric acid | |
| 12°C to 14°C | |
| 274°C to 275°C | |
| C6H10O3 | |
| MFCD00004412 | |
| 385840 | |
| Slightly soluble in water,chloroform and methanol. | |
| CC(=O)CCCC(=O)O | |
| 130.14 | |
| CHEBI:15888 | |
| 97% |
| 3128-06-1 | |
| 1.1 | |
| >110°C (230°F) | |
| 1.4451 | |
| 25 g | |
| 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid | |
| MGTZCLMLSSAXLD-UHFFFAOYSA-N | |
| 5-oxohexanoic acid | |
| 18407 | |
| 130.14 |
Chemical Identifiers
| 3128-06-1 | |
| 130.14 | |
| MGTZCLMLSSAXLD-UHFFFAOYSA-N | |
| CHEBI:15888 |
| C6H10O3 | |
| MFCD00004412 | |
| 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 221-511-7
TSCA : Yes
RUO – Research Use Only