missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbutyric acid, 97%

Catalog No. p-7044001
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0397806 5 g
AAL0397814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0397806 Supplier Thermo Scientific Chemicals Supplier No. L0397806
Only null left

CAS: 3128-06-1 | C6H10O3 | 130.14 g/mol

4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbutyric acid is used in the synthesis of selective indomethacin analogues for AKR1C3 inhibition in the treatment of castrate-resistant prostate cancer. It is also used to prepare PARP inhibitors for the treatment of cancer.

Solubility
Slightly soluble in water, chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3128-06-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00004412
InChI Key MGTZCLMLSSAXLD-UHFFFAOYSA-N
Synonym 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid

Specifications

Melting Point 12°C to 14°C
Density 1.1
Boiling Point 274°C to 275°C
Flash Point >110°C (230°F)
Refractive Index 1.4451
Quantity 5 g
Beilstein 385840
Solubility Information Slightly soluble in water,chloroform and methanol.
SMILES CC(=O)CCCCC(=O)O
IUPAC Name 6-oxoheptanoic acid
PubChem CID 98810
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material 4-Acetylbutyric acid
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 221-511-7
TSCA Yes

RUO – Research Use Only