missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzonitrile, 98+%

Catalog No. p-3737924
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167580050 5 g Glass bottle
AC167580250 25 g Glass bottle
AC167581000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167580050 Supplier Thermo Scientific Chemicals Supplier No. 167580050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1443-80-7 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1443-80-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001825
InChI Key NLPHXWGWBKZSJC-UHFFFAOYSA-N
Synonym 4'-cyanoacetophenone, benzonitrile, 4-acetyl, p-cyanoacetophenone, 4-cyanoacetophenone, p-acetylbenzonitrile, 4-acetyl-benzonitrile, benzonitrile, p-acetyl, 4-acetylbenzenecarbonitrile, 4-acetylbenznitrile
PubChem CID 74044
IUPAC Name 4-acetylbenzonitrile
SMILES CC(=O)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 55°C to 58°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H4CN
Quantity 5 g
Beilstein 10, 695
Formula Weight 145.16
Percent Purity 98+%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Chemical Name or Material 4-Acetylbenzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Obtain special instructions before use.
EINECSNumber 215-885-

RUO – Research Use Only