accessibility menu, dialog, popup

UserName

H3CON

CAS RN 1443-80-7

IUPAC Name: 4-acetylbenzonitrile
Synonyms: P-cyanoacetophenone
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: NLPHXWGWBKZSJC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)