missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylbenzoic Acid 98.0+%, TCI America™
$68.97 - $193.86
Chemical Identifiers
| CAS | 586-89-0 |
|---|---|
| Molecular Formula | C9H8O3 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00002561 |
| InChI Key | QBHDSQZASIBAAI-UHFFFAOYSA-N |
| Synonym | benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid |
| PubChem CID | 11470 |
| IUPAC Name | 4-acetylbenzoic acid |
| SMILES | CC(=O)C1=CC=C(C=C1)C(=O)O |
Chemical Identifiers
| 586-89-0 | |
| 164.16 | |
| QBHDSQZASIBAAI-UHFFFAOYSA-N | |
| 11470 | |
| CC(=O)C1=CC=C(C=C1)C(=O)O |
| C9H8O3 | |
| MFCD00002561 | |
| benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid | |
| 4-acetylbenzoic acid |
Specifications
| 586-89-0 | |
| White-Yellow | |
| MFCD00002561 | |
| benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid | |
| CC(=O)C1=CC=C(C=C1)C(=O)O | |
| 164.16 | |
| 164.16 | |
| Crystalline Powder |
| 210°C | |
| C9H8O3 | |
| 1 g | |
| QBHDSQZASIBAAI-UHFFFAOYSA-N | |
| 4-acetylbenzoic acid | |
| 11470 | |
| ≥98.0% (T) | |
| 4-Acetylbenzoic Acid |
Safety and Handling
TSCA : No