accessibility menu, dialog, popup

UserName

CAS RN 586-89-0

IUPAC Name: 4-acetylbenzoic acid
Synonyms: P-acetylbenzoic acid
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: QBHDSQZASIBAAI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)