missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzoic Acid 98.0+%, TCI America™

Catalog No. A10245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10245G 5 g
A10241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10245G Supplier TCI America Supplier No. A10245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 586-89-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002561
InChI Key QBHDSQZASIBAAI-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
PubChem CID 11470
IUPAC Name 4-acetylbenzoic acid
SMILES CC(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetylbenzoic Acid