missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzoic acid, 98+%

CAS: 586-89-0 | C9H8O3 | 164.16 g/mol

$73.26 - $947.97

Chemical Identifiers

CAS 586-89-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002561
InChI Key QBHDSQZASIBAAI-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
PubChem CID 11470
IUPAC Name 4-acetylbenzoic acid
SMILES CC(=O)C1=CC=C(C=C1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1523903
View Documents
Thermo Scientific Chemicals
A1523903
1 g
Each for $73.26
Only null left
 
AAA1523906
View Documents
Thermo Scientific Chemicals
A1523906
5 g
Each for $263.03
Only null left
 
AAA1523914
View Documents
Thermo Scientific Chemicals
A1523914
25 g
Each for $947.97
Only null left
 
Description

Description

4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

586-89-0
164.16
QBHDSQZASIBAAI-UHFFFAOYSA-N
11470
CC(=O)C1=CC=C(C=C1)C(=O)O
C9H8O3
MFCD00002561
benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
4-acetylbenzoic acid

Specifications

586-89-0
Odorless
MFCD00002561
2207355
Soluble in water.
CC(=O)C1=CC=C(C=C1)C(=O)O
164.16
164.16
4-Acetylbenzoic acid
208°C to 212°C
C9H8O3
1 g
benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
QBHDSQZASIBAAI-UHFFFAOYSA-N
4-acetylbenzoic acid
11470
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-588-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only