missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzoic acid, 98+%

Catalog No. AAA1523903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1523903 1 g
AAA1523906 5 g
AAA1523914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1523903 Supplier Thermo Scientific Chemicals Supplier No. A1523903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 586-89-0 | C9H8O3 | 164.16 g/mol

4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 586-89-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002561
InChI Key QBHDSQZASIBAAI-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
PubChem CID 11470
IUPAC Name 4-acetylbenzoic acid
SMILES CC(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 208°C to 212°C
Odor Odorless
Quantity 1 g
Beilstein 2207355
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material 4-Acetylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-588-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only