missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxystyrene, 96%, stabilized with 200-300 ppm MEHQ, Thermo Scientific Chemicals

Catalog No. p-3771095
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427000050 5 mL Glass bottle
AC427000250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427000050 Supplier Thermo Scientific Chemicals Supplier No. 427000050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2628-16-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00075734
InChI Key JAMNSIXSLVPNLC-UHFFFAOYSA-N
Synonym 4-acetoxystyrene, 4-vinylphenyl acetate, 4-ethenylphenol acetate, p-acetoxystyrene, phenol, 4-ethenyl-, acetate, p-vinylphenol acetate, unii-7s21904ayn, phenol, 4-ethenyl-, 1-acetate, 4-ethenylphenyl acetate, acetic acid 4-vinylphenyl ester
PubChem CID 75821
IUPAC Name (4-ethenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 7.0°C to 8.0°C
Color Colorless
Density 1.0600g/mL
Boiling Point 260.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3COOC6H4CH=CH2
Refractive Index 1.5375 to 1.5395 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.06
Solubility Information Solubility in water: 0.1g/L (20°C)
Stabilizer stabilized with 200-300ppm MeHQ
Formula Weight 162.19
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Acetoxystyrene, Stabilized with 200 to 300 ppm MEHQ
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice

RUO – Research Use Only