missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxystyrene, 95%, stab.

Catalog No. AAL1160109
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1160109 10 g
AAL1160104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1160109 Supplier Thermo Scientific Chemicals Supplier No. L1160109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2628-16-2 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2628-16-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00075734
InChI Key JAMNSIXSLVPNLC-UHFFFAOYSA-N
Synonym 4-acetoxystyrene, 4-vinylphenyl acetate, 4-ethenylphenol acetate, p-acetoxystyrene, phenol, 4-ethenyl-, acetate, p-vinylphenol acetate, unii-7s21904ayn, phenol, 4-ethenyl-, 1-acetate, 4-ethenylphenyl acetate, acetic acid 4-vinylphenyl ester
PubChem CID 75821
IUPAC Name (4-ethenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=C

Specifications

Melting Point 7°C to 8°C
Density 1.06
Boiling Point 259°C to 260°C
Flash Point 87°C (188°F)
Refractive Index 1.538
Quantity 10 g
Beilstein 1862793
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material 4-Acetoxystyrene, stabilized
Hazard Category H227-H302-H315-H317-H319-H334
Hazard Statement GHS H Statement
H334-H302-H227-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
EINECSNumber 434-600-2
RTECSNumber SL3784000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only