missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzonitrile, 97%, Thermo Scientific™

Catalog No. p-7036113
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5435803 1 g
AAH5435806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5435803 Supplier Thermo Scientific Chemicals Supplier No. H5435803
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Acetoxybenzonitrile is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13031-41-9
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00129158
InChI Key CJGXWABHYYJNJH-UHFFFAOYSA-N
Synonym p-cyanophenyl acetate, benzonitrile, 4-acetyloxy, 4-acetoxybenzonitrile, unii-eel1pcp8am, eel1pcp8am, 4-cyanophenyl acetate, 4-acetyloxy benzonitrile, benzonitrile, p-hydroxy-, acetate ester
PubChem CID 83062
IUPAC Name (4-cyanophenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C#N

Specifications

Quantity 1 g
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 4-Acetoxybenzonitrile
Hazard Category H302+H312-H317-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P333+P313-P363-P501c
EINECSNumber 235-893-8
TSCA Yes
Recommended Storage Ambient temperatures