missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117109
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088525G 25 g
A0885500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A088525G Supplier TCI America Supplier No. A088525G
Only null left

Chemical Identifiers

CAS 2345-34-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002540
InChI Key GDBUZIKSJGRBJP-UHFFFAOYSA-N
Synonym 4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid
PubChem CID 16865
ChEBI CHEBI:86560
IUPAC Name 4-(acetyloxy)benzoic acid
SMILES CC(=O)OC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 193°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetoxybenzoic Acid
EINECSNumber (3)-1652
TSCA Yes