missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7117457
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14675G 5 g
A146725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14675G Supplier TCI America Supplier No. A14675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 878-00-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00003384
InChI Key SEVSMVUOKAMPDO-UHFFFAOYSA-N
Synonym 4-acetoxybenzaldehyde, p-acetoxybenzaldehyde, benzaldehyde, 4-acetyloxy, p-hydroxybenzaldehyde acetate, 4-acetoxy benzaldehyde, 4-formylphenyl acetate, 4-acetyloxybenzaldehyde, acetic acid 4-formylphenyl ester, benzaldehyde, p-hydroxy-, acetate, benzaldehyde,4-acetyloxy
PubChem CID 70144
ChEBI CHEBI:86559
IUPAC Name (4-formylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 265°C
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Acetoxybenzaldehyde