Learn More
4-Acetamidophenol, 98%
CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol
Supplier: Thermo Scientific Chemicals 102330010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Acetamidophenol | |
| 97.5 | |
| 168.0°C to 172.0°C | |
| 1.2930g/mL | |
| 97.5% min. (HPLC) | |
| C8H9NO2 | |
| MFCD00002328 | |
| 13,46 | |
| 15,47 | |
| Solubility in water: soluble 14g/L in water (20°C). Other solubilities: soluble in ethanol,acetone,alcohol,methanol,,dmf,ethylene dichloride,ethyl acetate,practically insoluble in petr.ether,pentane and,benzene | |
| CC(=O)NC1=CC=C(O)C=C1 | |
| 151.17 | |
| CHEBI:46195 | |
| 98% |
| 103-90-2 | |
| 100.0 | |
| White | |
| Authentic | |
| Plastic bottle | |
| CH3CONHC6H4OH | |
| 1 kg | |
| 1.293 | |
| acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol | |
| RZVAJINKPMORJF-UHFFFAOYSA-N | |
| N-(4-hydroxyphenyl)acetamide | |
| 1983 | |
| 151.16 | |
| Crystalline Powder or Crystals |
Chemical Identifiers
| 103-90-2 | |
| 151.17 | |
| RZVAJINKPMORJF-UHFFFAOYSA-N | |
| 1983 | |
| N-(4-hydroxyphenyl)acetamide |
| C8H9NO2 | |
| MFCD00002328 | |
| acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol | |
| CHEBI:46195 | |
| CC(=O)NC1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this p
GHS Signal Word: Warning
EINECSNumber : 203-157-5
RUO – Research Use Only