missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidocyclohexanone 98.0+%, TCI America™

Catalog No. A19295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19295G 5 g
A192925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19295G Supplier TCI America Supplier No. A19295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27514-08-5
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.197
MDL Number MFCD03703462
InChI Key WZEMYWNHKFIVKE-UHFFFAOYSA-N
Synonym n-4-oxocyclohexyl acetamide, 4-acetamidocyclohexanone, 4-acetamido-cyclohexanone, 4-acetylamino cyclohexanone, 4-n-acetylamino cyclohexanone, 4-n-acetyl-amino-cyclohexanone, n-4-oxo-cyclohexyl-acetamide, acetamide, n-4-oxocyclohexyl, 4-aminocyclohexanone, n-acetyl
PubChem CID 538565
IUPAC Name N-(4-oxocyclohexyl)acetamide
SMILES CC(=O)NC1CCC(=O)CC1

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 5 g
Formula Weight 155.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidocyclohexanone