accessibility menu, dialog, popup

UserName

CH3ONHO

CAS RN 27514-08-5

IUPAC Name: N-(4-oxocyclohexyl)acetamide
Synonyms: N-(4-oxocyclohexyl)acetamide
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: WZEMYWNHKFIVKE-UHFFFAOYSA-N
SMILES: CC(=O)NC1CCC(=O)CC1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)