missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzoic Acid 98.0+%, TCI America™

Catalog No. A001325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A001325G 25 g
A0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A001325G Supplier TCI America Supplier No. A001325G

Chemical Identifiers

CAS 556-08-1
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00002534
InChI Key QCXJEYYXVJIFCE-UHFFFAOYSA-N
Synonym acedoben, p-acetamidobenzoic acid, 4-acetylamino benzoic acid, 4-carboxyacetanilide, 4-acetylaminobenzoic acid, benzoic acid, 4-acetylamino, p-acetylamino benzoic acid, p-acetoaminobenzoic acid, p-acetaminobenzoic acid, n-acetyl-p-aminobenzoic acid
PubChem CID 19266
ChEBI CHEBI:46171
IUPAC Name 4-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 262°C
Color White
Quantity 25 g
Formula Weight 179.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidobenzoic Acid
EINECSNumber (3)-1450
TSCA Yes