missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetamidobenzenesulfonyl Azide 98.0+%, TCI America™
Supplier: TCI America A178625G
Specifications
| 4-Acetamidobenzenesulfonyl Azide | |
| 112°C | |
| C8H8N4O3S | |
| 25 g | |
| NTMHWRHEGDRTPD-UHFFFAOYSA-N | |
| 4-acetamidobenzene-1-sulfonyl azide | |
| 5129185 | |
| ≥98.0% (HPLC) |
| 2158-14-7 | |
| Purple-White | |
| MFCD00029626 | |
| 4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide | |
| CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-] | |
| 240.24 | |
| 240.24 | |
| Crystalline Powder |
Chemical Identifiers
| 2158-14-7 | |
| 240.24 | |
| NTMHWRHEGDRTPD-UHFFFAOYSA-N | |
| 5129185 | |
| CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-] |
| C8H8N4O3S | |
| MFCD00029626 | |
| 4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide | |
| 4-acetamidobenzene-1-sulfonyl azide |
Safety and Handling
TSCA : No