missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzenesulfonyl azide, 97%

Catalog No. AAL1591606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1591606 5 g
AAL1591614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1591606 Supplier Thermo Scientific Chemicals Supplier No. L1591606

CAS: 2158-14-7 | C8H8N4O3S | 240.24 g/mol

4-Acetamidobenzenesulfonyl azide is used as a hydroazidation catalyst for facile preparation of organoazides. It is used as a reagent for synthesis of: monosaccharide-derived alcohols, a late-stage intermolecular C-H olefination, intramolecular isomuenchnone cycloaddition approach to antitumor agents, rhodium-catalyzed carbene cyclization cycloaddition cascade reaction of vinylsulfonates and Suzuki-Miyaura cross coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetamidobenzenesulfonyl azide is used as a hydroazidation catalyst for facile preparation of organoazides. It is used as a reagent for synthesis of: monosaccharide-derived alcohols, a late-stage intermolecular C-H olefination, intramolecular isomuenchnone cycloaddition approach to antitumor agents, rhodium-catalyzed carbene cyclization cycloaddition cascade reaction of vinylsulfonates and Suzuki-Miyaura cross coupling reaction.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2158-14-7
Molecular Formula C8H8N4O3S
Molecular Weight (g/mol) 240.24
MDL Number MFCD00029626
InChI Key NTMHWRHEGDRTPD-UHFFFAOYSA-N
Synonym 4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide
PubChem CID 5129185
IUPAC Name N-(4-azidosulfonylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]

Specifications

Melting Point 107°C to 111°C
Quantity 5 g
Beilstein 2219568
Solubility Information Insoluble in water.
Formula Weight 240.24
Percent Purity 97%
Chemical Name or Material 4-Acetamidobenzenesulfonyl azide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 606-801-7
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only