missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzenesulfonyl azide, 97%

Catalog No. p-7077303
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443800050 5 g
AC443800250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443800050 Supplier Thermo Scientific Chemicals Supplier No. 443800050
Only null left

CAS: 2158-14-7 | C8H8N4O3S | 240.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2158-14-7
Molecular Formula C8H8N4O3S
Molecular Weight (g/mol) 240.24
MDL Number MFCD00029626
InChI Key NTMHWRHEGDRTPD-UHFFFAOYSA-N
Synonym 4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide
PubChem CID 5129185
IUPAC Name N-(4-azidosulfonylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]

Specifications

Melting Point 107.0°C to 111.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 240.24
Percent Purity 97%
Chemical Name or Material 4-Acetamidobenzenesulfonyl azide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only