missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzeneboronic acid, 96%

Catalog No. p-7035273
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5298703 1 g
AAH5298706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5298703 Supplier Thermo Scientific Chemicals Supplier No. H5298703
Only null left

CAS: 101251-09-6 | C8H10BNO3 | 178.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101251-09-6
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
MDL Number MFCD02179451
InChI Key VYEWTHXZHHATTA-UHFFFAOYSA-N
Synonym 4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl
PubChem CID 2734657
IUPAC Name (4-acetamidophenyl)boronic acid
SMILES CC(=O)NC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 215°C to 220°C
Quantity 1 g
Formula Weight 178.98
Percent Purity 96%
Chemical Name or Material 4-Acetamidobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only