missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzaldehyde, 98%

Catalog No. AAB2043006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2043006 5 g
AAB2043014 25 g
AAB2043022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2043006 Supplier Thermo Scientific Chemicals Supplier No. B2043006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 122-85-0 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-85-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00003380
InChI Key SKLUWKYNZNXSLX-UHFFFAOYSA-N
Synonym 4-acetamidobenzaldehyde, n-4-formylphenyl acetamide, 4-acetylaminobenzaldehyde, p-formylacetanilide, acetamide, n-4-formylphenyl, micotiazone, 4-formylacetanilide, 4'-formylacetanilide, p-acetamidobenzaldehyde, p-acetaminobenzaldehyde
PubChem CID 73942
IUPAC Name N-(4-formylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C=O

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 4-Acetamidobenzaldehyde
EINECSNumber 204-579-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only