missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Acetamido-2'-methylacetophenone 98.0+%, TCI America™
$48.13 - $146.31
Chemical Identifiers
| CAS | 34956-31-5 |
|---|---|
| Molecular Formula | C11H13NO2 |
| Molecular Weight (g/mol) | 191.23 |
| MDL Number | MFCD02258876 |
| InChI Key | PTARWPNYVATTDE-UHFFFAOYSA-N |
| Synonym | n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl |
| PubChem CID | 2737381 |
| IUPAC Name | N-(4-acetyl-3-methylphenyl)acetamide |
| SMILES | CC(=O)NC1=CC(C)=C(C=C1)C(C)=O |
Chemical Identifiers
| 34956-31-5 | |
| 191.23 | |
| PTARWPNYVATTDE-UHFFFAOYSA-N | |
| 2737381 | |
| CC(=O)NC1=CC(C)=C(C=C1)C(C)=O |
| C11H13NO2 | |
| MFCD02258876 | |
| n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl | |
| N-(4-acetyl-3-methylphenyl)acetamide |
Specifications
| 34956-31-5 | |
| Yellow | |
| MFCD02258876 | |
| n-4-acetyl-3-methylphenyl acetamide, 4'-acetamido-2'-methylacetophenone, 4-acetamido-2-methylacetophenone, pubchem12562, 4'-acetyl-3'-methylacetoanilide, n-4-acetyl-3-methyl-phenyl-acetamide, 4\\'-acetamido-2\\'-methylacetophenone, n-4-ethanoyl-3-methyl-phenyl ethanamide, acetamide, n-4-acetyl-3-methylphenyl | |
| CC(=O)NC1=CC(C)=C(C=C1)C(C)=O | |
| 191.23 | |
| 191.23 | |
| Crystalline Powder |
| 139°C | |
| C11H13NO2 | |
| 5 g | |
| PTARWPNYVATTDE-UHFFFAOYSA-N | |
| N-(4-acetyl-3-methylphenyl)acetamide | |
| 2737381 | |
| ≥98.0% (GC) | |
| 4′-Acetamido-2′-methylacetophenone |
Safety and Handling
TSCA : No