missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-1-benzylpiperidine 98.0+%, TCI America™

Catalog No. A16545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16545G 5 g
A165425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16545G Supplier TCI America Supplier No. A16545G

Chemical Identifiers

CAS 50534-23-1
Molecular Formula C14H20N2O
Molecular Weight (g/mol) 232.33
MDL Number MFCD00247343
InChI Key PXKZVCOKJFOTQY-UHFFFAOYSA-N
PubChem CID 12641850
IUPAC Name N-(1-benzylpiperidin-4-yl)acetamide
SMILES CC(=O)NC1CCN(CC2=CC=CC=C2)CC1

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 232.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-1-benzylpiperidine