accessibility menu, dialog, popup

UserName

CH3ONHN

CAS RN 50534-23-1

IUPAC Name: N-(1-benzylpiperidin-4-yl)acetamide
Synonyms: N-(1-benzylpiperidin-4-yl)acetamide
Molecular Weight (g/mol): 232.33
Molecular Formula: C14H20N2O
InChi Key: PXKZVCOKJFOTQY-UHFFFAOYSA-N
SMILES: CC(=O)NC1CCN(CC2=CC=CC=C2)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results